Assay Details
| Gene Symbol: Atg16l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205391.1 | NP_001192320.1 | 6 - 7 | 894 | 74 | |
| NM_001205392.1 | NP_001192321.1 | 6 - 7 | 894 | 74 | ||
| NM_029846.4 | NP_084122.2 | 6 - 7 | 894 | 74 | ||
| GenBank mRNA | AB087879.1 | - | 6 - 7 | 705 | 74 | |
| AB087880.1 | - | 6 - 7 | 705 | 74 | ||
| AB087881.1 | - | 6 - 7 | 705 | 74 | ||
| AB476646.1 | - | 6 - 7 | 705 | 74 | ||
| AK005181.1 | - | 6 - 7 | 819 | 74 | ||
| AK030983.1 | - | 6 - 7 | 835 | 74 | ||
| AK131120.1 | - | 6 - 7 | 793 | 74 | ||
| AK155358.1 | - | 6 - 7 | 830 | 74 | ||
| AK163246.1 | - | 6 - 7 | 833 | 74 | ||
| AK170073.1 | - | 6 - 7 | 828 | 74 | ||
| AK218161.1 | - | 2 - 3 | 157 | 74 | ||
| BC049122.1 | - | 6 - 7 | 819 | 6813377 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atg16l1
Genomic Map