Assay Details
| Gene Symbol: Tmco4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029857.3 | NP_084133.1 | 9 - 10 | 1377 | 76 | |
| GenBank mRNA | AK014571.1 | - | 8 - 9 | 978 | 76 | |
| AK029092.1 | - | 9 - 10 | 1377 | 76 | ||
| AK170619.1 | - | 9 - 10 | 1106 | 76 | ||
| AK171189.1 | - | 9 - 10 | 1123 | 76 | ||
| BC013471.1 | - | 8 - 9 | 1003 | 4221072 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmco4
Genomic Map