Assay Details
| Gene Symbol: Tmco4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029857.3 | NP_084133.1 | 12 - 13 | 1804 | 66 | |
| GenBank mRNA | AK014571.1 | - | 11 - 12 | 1405 | 66 | |
| AK029092.1 | - | 12 - 13 | 1804 | 66 | ||
| AK054337.1 | - | 11 - 12 | 1633 | 66 | ||
| AK170619.1 | - | 12 - 13 | 1533 | 66 | ||
| AK171189.1 | - | 12 - 13 | 1550 | 66 | ||
| BC013471.1 | - | 11 - 12 | 1430 | 4221072 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmco4
Genomic Map