Assay Details
| Gene Symbol: Tmco4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029857.3 | NP_084133.1 | 14 - 15 | 2027 | 87 | |
| GenBank mRNA | AK014571.1 | - | 13 - 14 | 1628 | 87 | |
| AK029092.1 | - | 14 - 15 | 2027 | 87 | ||
| AK170619.1 | - | 14 - 15 | 1756 | 87 | ||
| AK171189.1 | - | 14 - 15 | 1773 | 87 | ||
More Information
Set Membership: |
|

Tmco4
Genomic Map