| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1500015726.mm.1 |
NM_029946.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015726.mm.1 |
NM_029946.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015726.mm.1 |
TC1500001981.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500015734.mm.1 |
NM_029946.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015734.mm.1 |
NM_029946.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015734.mm.1 |
TC1500001981.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500015736.mm.1 |
NM_029946.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015736.mm.1 |
NM_029946.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015736.mm.1 |
TC1500001981.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500015739.mm.1 |
NM_029946.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015739.mm.1 |
NM_029946.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015739.mm.1 |
TC1500001981.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500015723.mm.1 |
NM_029946.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015723.mm.1 |
NM_029946.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015723.mm.1 |
TC1500001981.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500015724.mm.1 |
NM_029946.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015724.mm.1 |
NM_029946.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015724.mm.1 |
TC1500001981.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500015738.mm.1 |
NM_029946.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015738.mm.1 |
NM_029946.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015738.mm.1 |
TC1500001981.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500015740.mm.1 |
NM_029946.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015740.mm.1 |
NM_029946.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015740.mm.1 |
TC1500001981.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500015714.mm.1 |
NM_001161628.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015714.mm.1 |
NM_001161628.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015714.mm.1 |
NM_029946.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015714.mm.1 |
NM_029946.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015714.mm.1 |
TC1500001981.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500015715.mm.1 |
NM_001161628.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015715.mm.1 |
NM_001161628.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015715.mm.1 |
TC1500001981.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500015732.mm.1 |
NM_029946.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015732.mm.1 |
NM_029946.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015732.mm.1 |
TC1500001981.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500015713.mm.1 |
NM_001161628.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015713.mm.1 |
NM_001161628.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015713.mm.1 |
NM_029946.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015713.mm.1 |
NM_029946.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015713.mm.1 |
TC1500001981.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500015719.mm.1 |
NM_029946.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015719.mm.1 |
NM_029946.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015719.mm.1 |
TC1500001981.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500015729.mm.1 |
NM_029946.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015729.mm.1 |
NM_029946.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015729.mm.1 |
TC1500001981.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500015735.mm.1 |
NM_029946.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015735.mm.1 |
NM_029946.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015735.mm.1 |
TC1500001981.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500015712.mm.1 |
NM_001161628.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015712.mm.1 |
NM_001161628.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015712.mm.1 |
NM_029946.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015712.mm.1 |
NM_029946.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015712.mm.1 |
TC1500001981.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500015720.mm.1 |
NM_029946.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015720.mm.1 |
NM_029946.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015720.mm.1 |
TC1500001981.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500015722.mm.1 |
NM_029946.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015722.mm.1 |
NM_029946.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015722.mm.1 |
TC1500001981.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500015731.mm.1 |
NM_029946.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015731.mm.1 |
NM_029946.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015731.mm.1 |
TC1500001981.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500015737.mm.1 |
NM_029946.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015737.mm.1 |
NM_029946.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015737.mm.1 |
TC1500001981.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500015711.mm.1 |
NM_001161628.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015711.mm.1 |
NM_001161628.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015711.mm.1 |
NM_029946.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015711.mm.1 |
NM_029946.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015711.mm.1 |
TC1500001981.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500015716.mm.1 |
NM_029946.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015716.mm.1 |
NM_029946.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015716.mm.1 |
TC1500001981.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500015718.mm.1 |
NM_029946.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015718.mm.1 |
NM_029946.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015718.mm.1 |
TC1500001981.mm.1 |
MTA TC |