Assay Details
| Gene Symbol: Lrrc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142728.1 | NP_001136200.1 | 8 - 9 | 1216 | 84 | |
| NM_001142729.1 | NP_001136201.1 | 8 - 9 | 1135 | 84 | ||
| NM_030070.3 | NP_084346.2 | 8 - 9 | 1216 | 84 | ||
| GenBank mRNA | AK015292.1 | - | 8 - 9 | 1133 | 84 | |
| AK019545.1 | - | 8 - 9 | 1149 | 84 | ||
| AK029802.1 | - | 8 - 9 | 1217 | 84 | ||
| AK031279.1 | - | 8 - 9 | 1134 | 84 | ||
More Information
Set Membership: |
|

Lrrc9
Genomic Map