Assay Details
| Gene Symbol: Clip4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271483.1 | NP_001258412.1 | 11 - 12 | 1539 | 68 | |
| NM_001271484.1 | NP_001258413.1 | 11 - 12 | 1624 | 68 | ||
| NM_030179.3 | NP_084455.2 | 11 - 12 | 1527 | 68 | ||
| NM_175378.1 | NP_780587.1 | 10 - 11 | 1483 | 68 | ||
| GenBank mRNA | AK017914.1 | - | 8 - 9 | 1115 | 68 | |
| AK019516.1 | - | 11 - 12 | 1540 | 68 | ||
| AK075758.1 | - | 10 - 11 | 1474 | 68 | ||
| AK161301.1 | - | 11 - 12 | 1614 | 68 | ||
| AK163644.1 | - | 4 - 5 | 467 | 68 | ||
| BC011280.1 | - | 4 - 5 | 391 | 68 | ||
| BC035226.1 | - | 11 - 12 | 1527 | 4920788 | 68 | |
More Information
Set Membership: |
|

Clip4
Genomic Map