Assay Details
| Gene Symbol: Usp54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030180.2 | NP_084456.2 | 18 - 19 | 4643 | 64 | |
| GenBank mRNA | AK019588.1 | - | 1 - 2 | 830 | 64 | |
| BC066166.1 | - | 1 - 2 | 1154 | 64 | ||
| BC066175.1 | - | 1 - 2 | 1154 | 64 | ||
| BC117844.1 | - | 1 - 2 | 818 | 64 | ||
| BC117845.1 | - | 1 - 2 | 818 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Usp54
Genomic Map