Assay Details
| Gene Symbol: Armc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027456.1 | NP_081732.1 | 14 - 15 | 1410 | 87 | |
| NM_030184.2 | NP_084460.1 | 14 - 15 | 1445 | 87 | ||
| GenBank mRNA | AK017564.1 | - | 14 - 15 | 1411 | 87 | |
| AK019600.1 | - | 14 - 15 | 1434 | 87 | ||
| AK030307.1 | - | 14 - 15 | 1442 | 87 | ||
| AK036083.1 | - | 14 - 15 | 1416 | 87 | ||
| AK039800.1 | - | 14 - 15 | 1418 | 87 | ||
| AK045699.1 | - | 14 - 15 | 1409 | 87 | ||
| AK046693.1 | - | 14 - 15 | 1428 | 87 | ||
| AK140598.1 | - | 14 - 15 | 1441 | 87 | ||
| AK161393.1 | - | 14 - 15 | 1425 | 87 | ||
| AK164041.1 | - | 14 - 15 | 1444 | 87 | ||
| BC028964.1 | - | 7 - 8 | 665 | 5364429 | 87 | |
| BC043101.1 | - | 14 - 15 | 1397 | 6406321 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Armc9
Genomic Map