Assay Details
| Gene Symbol: Stpg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030189.3 | NP_084465.1 | 4 - 5 | 511 | 78 | |
| GenBank mRNA | AK019766.1 | - | 4 - 5 | 512 | 78 | |
| BC145657.1 | - | 3 - 4 | 298 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stpg1
Genomic Map