Assay Details
| Gene Symbol: Stpg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030189.3 | NP_084465.1 | 6 - 7 | 798 | 78 | |
| GenBank mRNA | AK015061.1 | - | 7 - 8 | 992 | 78 | |
| AK019766.1 | - | 6 - 7 | 799 | 78 | ||
| BC145657.1 | - | 5 - 6 | 585 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stpg1
Genomic Map