Assay Details
| Gene Symbol: Stpg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030189.3 | NP_084465.1 | 7 - 8 | 959 | 79 | |
| GenBank mRNA | AK015061.1 | - | 8 - 9 | 79 | ||
| AK019766.1 | - | 7 - 8 | 960 | 79 | ||
| BC145657.1 | - | 6 - 7 | 746 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stpg1
Genomic Map