Assay Details
| Gene Symbol: Pagr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030240.1 | NP_084516.1 | 2 - 3 | 995 | 73 | |
| GenBank mRNA | AK013840.1 | - | 2 - 3 | 969 | 73 | |
| AK082775.1 | - | 2 - 3 | 704 | 73 | ||
| BC003932.1 | - | 2 - 3 | 995 | 3485982 | 73 | |
| BC054744.1 | - | 4132 | 73 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25503619 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25503618 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25503585 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25503589 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25503586 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25503620 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25503588 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P279623 | BC003932.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P279623 | BC054744.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P279623 | NM_030240.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P279623 | AK013840.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P279623 | BC003932.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P279623 | BC054744.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P279623 | NM_030240.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P279623 | AK013840.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 485572 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P174228 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P279623 | BC003932.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P279623 | BC054744.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P279623 | NM_030240.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P279623 | AK013840.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P279623 | BC003932.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P279623 | BC054744.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P279623 | NM_030240.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P279623 | AK013840.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503614 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503588 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503604 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503590 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503606 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503605 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503584 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503579 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503594 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503611 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503569 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503578 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503624 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503625 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503598 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503610 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503573 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503599 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503596 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503622 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503574 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503630 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503628 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503600 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503607 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503592 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503591 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503612 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503616 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503620 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503609 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503595 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503586 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503576 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503593 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503582 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503617 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503626 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503571 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503580 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503623 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503602 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25503618 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P174228 | - | - |
| Whole Mouse Genome G4122A | A_51_P279623 | BC003932.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P279623 | BC054744.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P279623 | NM_030240.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P279623 | AK013840.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P279623 | BC003932.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P279623 | BC054744.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P279623 | NM_030240.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P279623 | AK013840.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122920 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112104 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P174228 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05962042 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06030403 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06144115 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06144112 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05962032 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06030399 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06144113 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05962028 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25503619 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25503589 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25503586 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25503588 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25503585 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25503618 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25503620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25503524 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06144115 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25503588 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25503578 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05962026 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05962031 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25503580 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05962032 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25503573 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25503569 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25503576 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25503574 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10568136 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568138 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568133 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568134 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568137 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568132 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424014_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449618_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449617_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10568135 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10568131 | - | - |
| Mouse Genome 430 2.0 Array | 1429113_at | - | - |
| Mouse Genome 430 2.0 Array | 1424014_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1424014_at | - | - |
| Mouse Genome 430 2.0 Array | 1449617_at | - | - |
| Mouse Genome 430 2.0 Array | 1449617_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1449618_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1429113_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1449618_PM_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67278_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67278_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700039196.mm.1 | NM_030240.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700039196.mm.1 | NM_030240.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700039196.mm.1 | TC0700004661.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1424014_PM_at | - | - |
| Mouse Expression Set 430 | 1424014_at | - | - |
| Mouse Expression Set 430 | 1429113_at | - | - |
| Mouse Expression Set 430 | 1429113_PM_at | - | - |
| Murine Genome U74v2 Set | 162165_f_at | - | - |
| Murine Genome U74v2 Set | 95759_at | - | - |
| Murine Genome U74v2 Set | 95759_PM_at | - | - |
| Murine Genome U74v2 Set | 162165_PM_f_at | - | - |
| Murine Genome U74v2 Set | 169627_at | - | - |
| Murine Genome U74v2 Set | 169627_PM_at | - | - |
| Murine Genome U74v2 Set | 169105_r_at | - | - |
| Murine Genome U74v2 Set | 170128_PM_i_at | - | - |
| Murine Genome U74v2 Set | 169105_PM_r_at | - | - |
| Murine Genome U74v2 Set | 170128_i_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424014_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424014_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449618_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449618_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424014_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449617_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449617_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424014_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 542987 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1770435 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001770435 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2610992 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2610992 | - | - |
| mouse-6 v1.1 (Illumina) | scl067278.1_10-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1770435 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl067278.1_10-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1770435 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl067278.1_10-S | - | - |

Pagr1a
Genomic Map