Assay Details
| Gene Symbol: Wwtr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168281.1 | NP_001161753.1 | 4 - 5 | 965 | 86 | |
| NM_133784.3 | NP_598545.2 | 4 - 5 | 1044 | 86 | ||
| GenBank mRNA | AJ299430.1 | - | 4 - 5 | 979 | 86 | |
| AK011504.1 | - | 4 - 5 | 1018 | 86 | ||
| AK142299.1 | - | 4 - 5 | 966 | 86 | ||
| AK142720.1 | - | 4 - 5 | 969 | 86 | ||
| BC004640.1 | - | 4 - 5 | 965 | 3157246 | 86 | |
| BC014727.1 | - | 4 - 5 | 996 | 4487316 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wwtr1
Genomic Map