Assay Details
| Gene Symbol: Fgg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133862.1 | NP_598623.1 | 5 - 6 | 544 | 71 | |
| GenBank mRNA | AK149375.1 | - | 5 - 6 | 580 | 71 | |
| AK149448.1 | - | 5 - 6 | 581 | 71 | ||
| BC019506.1 | - | 5 - 6 | 544 | 4193631 | 71 | |
| BC019828.1 | - | 5 - 6 | 554 | 5123696 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fgg
Genomic Map