Assay Details
| Gene Symbol: Rcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197082.1 | NP_001184011.1 | 8 - 9 | 971 | 116 | |
| NM_133878.3 | NP_598639.1 | 7 - 8 | 932 | 116 | ||
| GenBank mRNA | AK076089.1 | - | 7 - 8 | 933 | 116 | |
| AK150153.1 | - | 7 - 8 | 884 | 116 | ||
| AK150610.1 | - | 7 - 8 | 882 | 116 | ||
| AK153193.1 | - | 7 - 8 | 882 | 116 | ||
| AK155225.1 | - | 5 - 6 | 514 | 116 | ||
| BC019807.1 | - | 7 - 8 | 844 | 4456795 | 116 | |
More Information
Set Membership: |
|

Rcc1
Genomic Map