Assay Details
| Gene Symbol: Rcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197082.1 | NP_001184011.1 | 9 - 10 | 1089 | 55 | |
| NM_133878.3 | NP_598639.1 | 8 - 9 | 1050 | 55 | ||
| GenBank mRNA | AK076089.1 | - | 8 - 9 | 1051 | 55 | |
| AK150153.1 | - | 8 - 9 | 1002 | 55 | ||
| AK150610.1 | - | 8 - 9 | 1000 | 55 | ||
| AK153193.1 | - | 8 - 9 | 1000 | 55 | ||
| AK155225.1 | - | 6 - 7 | 632 | 55 | ||
| BC019807.1 | - | 8 - 9 | 962 | 4456795 | 55 | |
| BC057645.1 | - | 9 - 10 | 1044 | 3591859 | 55 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rcc1
Genomic Map