Assay Details
| Gene Symbol: Rbm28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133925.2 | NP_598686.2 | 6 - 7 | 729 | 75 | |
| GenBank mRNA | AK013047.1 | - | 6 - 7 | 699 | 75 | |
| AK019296.1 | - | 6 - 7 | 678 | 75 | ||
| AK156773.1 | - | 6 - 7 | 666 | 75 | ||
| AK161670.1 | - | 6 - 7 | 695 | 75 | ||
| AK167050.1 | - | 6 - 7 | 730 | 75 | ||
| BC018373.1 | - | 2 - 3 | 103 | 3708613 | 75 | |
| BC024602.1 | - | 6 - 7 | 610 | 75 | ||
| BC040811.1 | - | 6 - 7 | 674 | 4015830 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rbm28
Genomic Map