Assay Details
| Gene Symbol: Rbm28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133925.2 | NP_598686.2 | 8 - 9 | 1032 | 78 | |
| GenBank mRNA | AK156773.1 | - | 8 - 9 | 969 | 78 | |
| AK161670.1 | - | 8 - 9 | 998 | 78 | ||
| AK167050.1 | - | 8 - 9 | 1034 | 78 | ||
| BC018373.1 | - | 4 - 5 | 406 | 3708613 | 78 | |
| BC024602.1 | - | 8 - 9 | 913 | 78 | ||
| BC040811.1 | - | 8 - 9 | 977 | 4015830 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rbm28
Genomic Map