Assay Details
| Gene Symbol: Ddx41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134059.2 | NP_598820.2 | 12 - 13 | 1451 | 63 | |
| GenBank mRNA | AK151427.1 | - | 12 - 13 | 1325 | 63 | |
| AK156957.1 | - | 12 - 13 | 1319 | 63 | ||
| BC011308.1 | - | 12 - 13 | 1305 | 3256762 | 63 | |
| BC023711.1 | - | 12 - 13 | 1326 | 63 | ||
| BE864921.1 | - | 3 - 4 | 242 | 63 | ||
| CX231996.1 | - | 5 - 6 | 644 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ddx41
Genomic Map