Assay Details
| Gene Symbol: Ddx41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134059.2 | NP_598820.2 | 15 - 16 | 1772 | 66 | |
| GenBank mRNA | AK151427.1 | - | 15 - 16 | 1646 | 66 | |
| AK156957.1 | - | 15 - 16 | 1640 | 66 | ||
| AK168043.1 | - | 15 - 16 | 1645 | 66 | ||
| AK180936.1 | - | 1 - 2 | 58 | 66 | ||
| BC011308.1 | - | 15 - 16 | 1626 | 3256762 | 66 | |
| BC023711.1 | - | 15 - 16 | 1647 | 66 | ||
| BE864921.1 | - | 6 - 7 | 563 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ddx41
Genomic Map