Assay Details
| Gene Symbol: Cdipt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026638.3 | NP_080914.1 | 1 - 2 | 895 | 85 | |
| GenBank mRNA | AK003041.1 | - | 1 - 2 | 446 | 85 | |
| AK076974.1 | - | 1 - 2 | 895 | 85 | ||
| AK079226.1 | - | 1 - 2 | 372 | 85 | ||
| AK089109.1 | - | 1 - 2 | 461 | 85 | ||
| AK132286.1 | - | 1 - 2 | 464 | 85 | ||
| BC021473.1 | - | 1 - 2 | 423 | 5343411 | 85 | |
| BC024413.1 | - | 1 - 2 | 235 | 4973307 | 85 | |
| BC032182.1 | - | 1 - 2 | 383 | 5252470 | 85 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK076974.1,AK079226.1,AK089109.1,BC021473.1,BC024413.1,BC032182.1,NM_026638.3 transcripts.
Set Membership: |
|

Cdipt
Genomic Map