Assay Details
| Gene Symbol: Arhgap17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122640.1 | NP_001116112.1 | 12 - 13 | 1156 | 113 | |
| NM_001122641.1 | NP_001116113.1 | 12 - 13 | 1156 | 113 | ||
| NM_001122642.1 | NP_001116114.1 | 12 - 13 | 1156 | 113 | ||
| NM_001122643.1 | NP_001116115.1 | 12 - 13 | 1156 | 113 | ||
| NM_144529.2 | NP_653112.2 | 12 - 13 | 1156 | 113 | ||
| GenBank mRNA | AB060553.1 | - | 12 - 13 | 1154 | 113 | |
| AB060554.1 | - | 12 - 13 | 1154 | 113 | ||
| AK036468.1 | - | 12 - 13 | 1093 | 113 | ||
| AK036617.1 | - | 12 - 13 | 1093 | 113 | ||
| AK046411.1 | - | 11 - 12 | 1063 | 113 | ||
| AK147010.1 | - | 12 - 13 | 1127 | 113 | ||
| AK149986.1 | - | 12 - 13 | 1128 | 113 | ||
| AK152266.1 | - | 12 - 13 | 1093 | 113 | ||
| AK152301.1 | - | 12 - 13 | 1120 | 113 | ||
| AK161636.1 | - | 12 - 13 | 1157 | 113 | ||
| BC003259.1 | - | 12 - 13 | 1104 | 3500822 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap17
Genomic Map