Assay Details
| Gene Symbol: Nmnat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144533.2 | NP_653116.1 | 3 - 4 | 731 | 71 | |
| GenBank mRNA | AK041526.1 | - | 3 - 4 | 460 | 71 | |
| AK041897.1 | - | 2 - 3 | 506 | 71 | ||
| BC005737.1 | - | 3 - 4 | 731 | 3599134 | 71 | |
| BC092086.1 | - | 3 - 4 | 440 | 5040019 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nmnat3
Genomic Map