Assay Details
| Gene Symbol: Ap5m1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144535.4 | NP_653118.3 | 2 - 3 | 1014 | 70 | |
| GenBank mRNA | AK016540.1 | - | 2 - 3 | 961 | 70 | |
| AK030928.1 | - | 2 - 3 | 941 | 70 | ||
| AK031527.1 | - | 2 - 3 | 1413 | 70 | ||
| AK051372.1 | - | 2 - 3 | 961 | 70 | ||
| AK154166.1 | - | 2 - 3 | 967 | 70 | ||
| AK164148.1 | - | 2 - 3 | 785 | 70 | ||
| AK170232.1 | - | 2 - 3 | 791 | 70 | ||
| BC023094.1 | - | 2 - 3 | 1014 | 4017596 | 70 | |
More Information
Set Membership: |
|

Ap5m1
Genomic Map