Assay Details
| Gene Symbol: Ap5m1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144535.4 | NP_653118.3 | 5 - 6 | 1474 | 129 | |
| GenBank mRNA | AK030928.1 | - | 5 - 6 | 1401 | 129 | |
| AK041400.1 | - | 5 - 6 | 975 | 129 | ||
| AK154166.1 | - | 5 - 6 | 1427 | 129 | ||
| AK164148.1 | - | 5 - 6 | 1245 | 129 | ||
| AK170232.1 | - | 5 - 6 | 1251 | 129 | ||
| AK205351.1 | - | 2 - 3 | 135 | 129 | ||
| AK211028.1 | - | 2 - 3 | 127 | 129 | ||
| BC023094.1 | - | 5 - 6 | 1474 | 4017596 | 129 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ap5m1
Genomic Map