Assay Details
| Gene Symbol: Ap5m1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144535.4 | NP_653118.3 | 6 - 7 | 1587 | 97 | |
| GenBank mRNA | AK016540.1 | - | 6 - 7 | 1536 | 97 | |
| AK030928.1 | - | 6 - 7 | 1514 | 97 | ||
| AK041400.1 | - | 6 - 7 | 1088 | 97 | ||
| AK154166.1 | - | 6 - 7 | 1540 | 97 | ||
| AK164148.1 | - | 6 - 7 | 1358 | 97 | ||
| AK170232.1 | - | 6 - 7 | 1364 | 97 | ||
| AK205351.1 | - | 3 - 4 | 248 | 97 | ||
| AK211028.1 | - | 3 - 4 | 240 | 97 | ||
| BC023094.1 | - | 6 - 7 | 1587 | 4017596 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ap5m1
Genomic Map