Assay Details
| Gene Symbol: Esco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC008220.1 | - | 6 - 7 | 854 | 117 | |
| BC172153.1 | - | 8 - 9 | 3060 | 117 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Esco1| Gene Symbol: Esco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC008220.1 | - | 6 - 7 | 854 | 117 | |
| BC172153.1 | - | 8 - 9 | 3060 | 117 | ||
Set Membership: |
|