Assay Details
| Gene Symbol: Nae1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144931.3 | NP_659180.1 | 1 - 2 | 127 | 78 | |
| GenBank mRNA | AK146202.1 | - | 1 - 2 | 85 | 78 | |
| AK162251.1 | - | 1 - 2 | 76 | 78 | ||
| AK166792.1 | - | 1 - 2 | 79 | 78 | ||
| BC019163.1 | - | 1 - 2 | 107 | 3711115 | 78 | |
| BC021510.1 | - | 1 - 2 | 108 | 5347277 | 78 | |
| BC023680.1 | - | 1 - 2 | 109 | 5343341 | 78 | |
| BC023897.1 | - | 1 - 2 | 84 | 5321187 | 78 | |
| BC027124.1 | - | 1 - 2 | 71 | 5355631 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nae1
Genomic Map