Assay Details
| Gene Symbol: Nae1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144931.3 | NP_659180.1 | 3 - 4 | 286 | 71 | |
| GenBank mRNA | AK146202.1 | - | 3 - 4 | 244 | 71 | |
| AK162251.1 | - | 3 - 4 | 235 | 71 | ||
| BC019163.1 | - | 3 - 4 | 266 | 3711115 | 71 | |
| BC021510.1 | - | 3 - 4 | 267 | 5347277 | 71 | |
| BC023680.1 | - | 3 - 4 | 268 | 5343341 | 71 | |
| BC023897.1 | - | 3 - 4 | 243 | 5321187 | 71 | |
| BC027124.1 | - | 3 - 4 | 230 | 5355631 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nae1
Genomic Map