Assay Details
| Gene Symbol: Nae1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144931.3 | NP_659180.1 | 16 - 17 | 1309 | 99 | |
| GenBank mRNA | AK146202.1 | - | 16 - 17 | 1267 | 99 | |
| BC019163.1 | - | 16 - 17 | 1289 | 3711115 | 99 | |
| BC021510.1 | - | 16 - 17 | 1290 | 5347277 | 99 | |
| BC023680.1 | - | 16 - 17 | 1291 | 5343341 | 99 | |
| BC023897.1 | - | 16 - 17 | 1266 | 5321187 | 99 | |
| BC027124.1 | - | 16 - 17 | 1253 | 5355631 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nae1
Genomic Map