Assay Details
| Gene Symbol: Prr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282012.1 | NP_001268941.1 | 2 - 3 | 374 | 103 | |
| NM_001282013.1 | NP_001268942.1 | 4 - 5 | 379 | 103 | ||
| NM_001282024.1 | NP_001268953.1 | 3 - 4 | 473 | 103 | ||
| NM_145487.3 | NP_663462.2 | 2 - 3 | 402 | 103 | ||
| GenBank mRNA | AJ504719.1 | - | 2 - 3 | 1171 | 103 | |
| AK132945.1 | - | 2 - 3 | 102 | 103 | ||
| AK218162.1 | - | 2 - 3 | 101 | 103 | ||
| BC016636.1 | - | 2 - 3 | 323 | 2650633 | 103 | |
| BU148736.1 | - | 4 - 5 | 336 | 103 | ||
| BY774318.1 | - | 3 - 4 | 301 | 103 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK132945.1,BC016636.1,NM_001282012.1,NM_001282013.1,NM_001282024.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31162041 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31162046 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31162043 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31162040 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31162039 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31162042 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31162044 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2161775 | AJ504719.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2161775 | NM_145487.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2161775 | NR_028507.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2161775 | NR_028516.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 470101 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 166059 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 31641 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 893131 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 84173 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P100748 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2161775 | AJ504719.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2161775 | NM_145487.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2161775 | NR_028507.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2161775 | NR_028516.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31162018 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31162011 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31162014 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31162022 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31162036 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31162019 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31162016 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31162023 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31162029 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31162041 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31162021 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31162038 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31162031 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31162012 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31162039 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31162040 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31162034 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31162044 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31162032 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31162026 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31162043 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08769738 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P100748 | - | - |
| Whole Mouse Genome G4122A | A_51_P498340 | BC016636.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P391825 | BP772693.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P391825 | AK016009.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P391825 | NM_001165892.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109265 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08769738 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P100748 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13224912 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13224907 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13224903 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13224899 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13224914 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31162043 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31162041 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31162039 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31162044 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31162040 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31162042 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31162046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31162082 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162054 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13255890 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162073 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13168383 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162094 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13100349 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162058 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162061 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162053 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13255888 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162044 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162070 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162071 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162040 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162047 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162099 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13168388 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162068 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162102 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13451852 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162074 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162096 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162100 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162092 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162043 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13451307 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162083 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13451316 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162066 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13322675 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162105 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13168389 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162050 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162085 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13089486 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162051 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162062 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13451304 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13285784 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162059 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162049 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13451850 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162104 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31162078 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10450671 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450670 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450674 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450672 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424145_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10450669 | - | - |
| Mouse Genome 430 2.0 Array | 1424145_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1424145_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75210_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75210_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700019035.mm.1 | NM_001282012.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019035.mm.1 | NM_001282013.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019035.mm.1 | NM_001282024.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019035.mm.1 | NM_145487.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019035.mm.1 | NM_145487.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019035.mm.1 | TC1700001991.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700019036.mm.1 | NM_001282012.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019036.mm.1 | NM_001282013.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019036.mm.1 | NM_001282024.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019036.mm.1 | NM_145487.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019036.mm.1 | NM_145487.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019036.mm.1 | TC1700001991.mm.1 | MTA TC |
| Affymetrix HT Mouse Genome 430A Array | 1424145_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424145_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 436484 | - | - |
| ABI Mouse Genome Survey Microarray | 608848 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3440112 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3440112 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003440112 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2716365 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2716365 | - | - |
| mouse-6 v1.1 (Illumina) | scl49968.5_292-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3440112 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl49968.5_292-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3440112 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2797322 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2797322 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl49968.5_292-S | - | - |

Prr3
Genomic Map