Assay Details
| Gene Symbol: Cald1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145575.3 | NP_663550.1 | 2 - 3 | 403 | 60 | |
| GenBank mRNA | AK160593.1 | - | 2 - 3 | 315 | 60 | |
| BC019435.1 | - | 2 - 3 | 362 | 5148205 | 60 | |
| BC055908.1 | - | 4 - 5 | 476 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cald1
Genomic Map