Assay Details
| Gene Symbol: Cald1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145575.3 | NP_663550.1 | 9 - 10 | 1615 | 60 | |
| GenBank mRNA | AK148009.1 | - | 7 - 8 | 987 | 60 | |
| BC019435.1 | - | 9 - 10 | 1574 | 5148205 | 60 | |
| CX568419.1 | - | 4 - 5 | 311 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cald1
Genomic Map