Assay Details
| Gene Symbol: Ranbp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145824.4 | NP_665823.2 | 2 - 3 | 515 | 89 | |
| GenBank mRNA | AK048772.1 | - | 2 - 3 | 371 | 89 | |
| AK173177.1 | - | 2 - 3 | 328 | 89 | ||
| AY337314.1 | - | 2 - 3 | 516 | 89 | ||
| BC024698.1 | - | 2 - 3 | 360 | 3667000 | 89 | |
| EU281316.1 | - | 2 - 3 | 376 | 89 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC024698.1 transcripts.
Set Membership: |
|

Ranbp10
Genomic Map