Assay Details
| Gene Symbol: Ranbp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145824.4 | NP_665823.2 | 8 - 9 | 1167 | 66 | |
| GenBank mRNA | AK173177.1 | - | 8 - 9 | 980 | 66 | |
| AY337314.1 | - | 8 - 9 | 1168 | 66 | ||
| BC024698.1 | - | 8 - 9 | 1012 | 3667000 | 66 | |
| EU281316.1 | - | 8 - 9 | 1028 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ranbp10
Genomic Map