Assay Details
| Gene Symbol: Tagap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145968.2 | NP_666080.1 | 7 - 8 | 749 | 76 | |
| GenBank mRNA | BC030886.1 | - | 7 - 8 | 729 | 4483459 | 76 |
| BC039780.1 | - | 7 - 8 | 740 | 4483459 | 76 | |
| BC150728.1 | - | 7 - 8 | 694 | 9087639 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tagap
Genomic Map