Assay Details
| Gene Symbol: Fam110a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146127.3 | NP_666239.1 | 1 - 2 | 228 | 66 | |
| GenBank mRNA | AK017381.1 | - | 1 - 2 | 122 | 66 | |
| AK050928.1 | - | 1 - 2 | 182 | 66 | ||
| BC034240.1 | - | 1 - 2 | 138 | 4208017 | 66 | |
| BC094463.1 | - | 1 - 2 | 163 | 4224947 | 66 | |
| BY119831.1 | - | 1 - 2 | 228 | 66 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK050928.1,BC034240.1,BC094463.1,NM_146127.3 transcripts.
Set Membership: |
|

Fam110a
Genomic Map