Assay Details
| Gene Symbol: Plekha4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148927.2 | NP_683729.2 | 7 - 8 | 811 | 64 | |
| GenBank mRNA | BC021387.1 | - | 7 - 8 | 822 | 5055226 | 64 |
| BC024961.1 | - | 5 - 6 | 770 | 4945500 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Plekha4
Genomic Map