Assay Details
| Gene Symbol: Plekha4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148927.2 | NP_683729.2 | 11 - 12 | 1308 | 71 | |
| GenBank mRNA | BC021387.1 | - | 11 - 12 | 1319 | 5055226 | 71 |
| BC024961.1 | - | 9 - 10 | 1267 | 4945500 | 71 | |
| BC030731.1 | - | 5 - 6 | 512 | 71 | ||
| BG081030.2 | - | 1 - 2 | 73 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plekha4
Genomic Map