Assay Details
Gene Symbol: Prss44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148940.3 | NP_683742.2 | 4 - 5 | 927 | 99 | |
GenBank mRNA | AB047758.1 | - | 861 | 99 | ||
AB162857.1 | - | 4 - 5 | 913 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2055112 | AB047758.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P11513 | NM_148940.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P11513 | AB162857.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 242573 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 825740 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 967646 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P114423 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P149875 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P11513 | NM_148940.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P11513 | AB162857.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826014 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26825991 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826008 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26825986 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26825994 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26825988 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826011 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826058 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26825996 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826002 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826071 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26825992 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826017 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826003 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826004 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826062 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826019 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826056 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826001 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826012 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26825997 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826064 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826006 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826061 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826069 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826066 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826005 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826010 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26826072 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26825993 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P149875 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02029690 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06926188 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P114423 | - | - |
Whole Mouse Genome G4122A | A_52_P181672 | NM_148940.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P181672 | AB047758.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P181672 | AB047759.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P181672 | AB162857.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133898 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P114423 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P149875 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08119710 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07999184 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07999180 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826075 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825977 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826069 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826026 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826058 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826022 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826001 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826061 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826078 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825993 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825986 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826071 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826006 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826024 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826019 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826003 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826089 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826017 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826054 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826027 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826005 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825997 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826056 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826084 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826081 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826029 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826008 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826062 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826087 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825991 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826012 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825988 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825996 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826021 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26825982 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826013 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826085 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826010 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26826004 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26826008 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26826006 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07754849 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26825991 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26826013 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26826003 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26826022 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26826027 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07482640 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26826010 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26826021 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26825997 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26825993 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26826005 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07868817 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26826029 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07754533 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26826004 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26826017 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07482634 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26825986 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26826012 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26825988 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26825979 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26825982 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07868814 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26826024 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26825977 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26825992 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26826019 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08119710 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26825978 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26825976 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07999184 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10589623 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10589619 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10589618 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10589622 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10589617 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10589621 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10589620 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1429995_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1427716_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1426131_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1447729_PM_s_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10589616 | - | - |
Mouse Genome 430 2.0 Array | 1447729_PM_s_at | - | - |
Mouse Genome 430 2.0 Array | 1426131_at | - | - |
Mouse Genome 430 2.0 Array | 1429995_at | - | - |
Mouse Genome 430 2.0 Array | 1429995_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1427716_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1426131_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1427716_at | - | - |
Mouse Genome 430 2.0 Array | 1447729_s_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73336_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73336_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0900012305.mm.1 | NM_148940.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900012305.mm.1 | NM_148940.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900012305.mm.1 | TC0900001470.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0900012307.mm.1 | NM_148940.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900012307.mm.1 | NM_148940.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900012307.mm.1 | TC0900001470.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0900012304.mm.1 | NM_148940.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900012304.mm.1 | NM_148940.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900012304.mm.1 | TC0900001470.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0900012306.mm.1 | NM_148940.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900012306.mm.1 | NM_148940.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900012306.mm.1 | TC0900001470.mm.1 | MTA TC |
Mouse Expression Set 430 | 1426131_at | - | - |
Mouse Expression Set 430 | 1427716_at | - | - |
Mouse Expression Set 430 | 1426131_PM_at | - | - |
Mouse Expression Set 430 | 1427716_PM_at | - | - |
Mouse Expression Set 430 | 1447729_PM_s_at | - | - |
Mouse Expression Set 430 | 1447729_s_at | - | - |
Mouse Expression Set 430 | 1429995_at | - | - |
Mouse Expression Set 430 | 1429995_PM_at | - | - |
Murine Genome U74v2 Set | 166931_PM_f_at | - | - |
Murine Genome U74v2 Set | 166931_f_at | - | - |
Mouse Genome 430A 2.0 Array | 1426131_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1427716_at | - | - |
Mouse Genome 430A 2.0 Array | 1427716_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1426131_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1427716_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1426131_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1427716_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1426131_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1429995_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1447729_s_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1447729_PM_s_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1429995_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 508541 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 3060010 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 3060010 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 102060746 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003060010 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1242978 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1242978 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1254770 | - | - |
mouse-6 v1.1 (Illumina) | scl36421.5.1_91-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 102060746 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3060010 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl36421.5.1_91-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102060746 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3060010 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2837999 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1242978 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2837999 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl36421.5.1_91-S | - | - |
