Assay Details
| Gene Symbol: Osbpl10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148958.2 | NP_683761.1 | 7 - 8 | 1425 | 69 | |
| GenBank mRNA | AK017034.1 | - | 9 - 10 | 1756 | 69 | |
| BC027256.1 | - | 7 - 8 | 1425 | 2650570 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Osbpl10
Genomic Map