Assay Details
| Gene Symbol: Osbpl10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148958.2 | NP_683761.1 | 8 - 9 | 1604 | 76 | |
| GenBank mRNA | AK017034.1 | - | 10 - 11 | 1935 | 76 | |
| BC027256.1 | - | 8 - 9 | 1604 | 2650570 | 76 | |
More Information
Set Membership: |
|

Osbpl10
Genomic Map