Assay Details
| Gene Symbol: Ugt2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152811.1 | NP_690024.1 | 1 - 2 | 725 | 111 | |
| GenBank mRNA | AK004971.1 | - | 1 - 2 | 752 | 111 | |
| AK050435.1 | - | 1 - 2 | 753 | 111 | ||
| BC027200.1 | - | 1 - 2 | 725 | 4161686 | 111 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P245503 | BC027200.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P245503 | AK004971.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P245503 | AK050435.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P245503 | NM_152811.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P245503 | BC027200.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P245503 | AK004971.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P245503 | AK050435.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P245503 | NM_152811.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 175323 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 363273 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 614295 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 620548 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 142200 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 1600 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P107196 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P178022 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P116995 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P245503 | BC027200.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P245503 | AK004971.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P245503 | AK050435.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P245503 | NM_152811.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P245503 | BC027200.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P245503 | AK004971.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P245503 | AK050435.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P245503 | NM_152811.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P245503 | BC027200.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P245503 | AK004971.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P245503 | AK050435.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P245503 | NM_152811.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P245503 | BC027200.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P245503 | AK004971.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P245503 | AK050435.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P245503 | NM_152811.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23865982 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23865984 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23865975 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23865981 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23865986 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23865995 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23865978 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23865983 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23865980 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23865997 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23865976 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23865989 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05643985 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P116995 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P178022 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P107196 | - | - |
| Whole Mouse Genome G4122A | A_51_P245503 | BC027200.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P245503 | AK004971.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P245503 | AK050435.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P245503 | NM_152811.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P245503 | BC027200.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P245503 | AK004971.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P245503 | AK050435.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P245503 | NM_152811.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134840 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124288 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P116995 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P178022 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P107196 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04783743 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04783746 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04783736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23866004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23865976 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23865941 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23865978 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04367574 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23865964 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23865981 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23865997 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23865960 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23865992 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23865953 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23865971 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23865986 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23865982 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23865947 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23865969 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23865980 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23865976 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23865945 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23865970 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23865942 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23865984 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23865955 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23865957 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23865949 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23865967 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23865954 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23865974 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23865951 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23865989 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04886436 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23865968 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04234961 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23865995 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23865961 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10531045 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10531043 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10531047 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10531046 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10531044 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10531042 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424934_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10531041 | - | - |
| Mouse Genome 430 2.0 Array | 1424934_at | - | - |
| Mouse Genome 430 2.0 Array | 1424934_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71773_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71773_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500023749.mm.1 | TC0500002690.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500023749.mm.1 | NM_152811.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500023749.mm.1 | NM_152811.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500023750.mm.1 | TC0500002690.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500023750.mm.1 | NM_152811.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500023750.mm.1 | NM_152811.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500023746.mm.1 | TC0500002690.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500023746.mm.1 | NM_152811.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500023746.mm.1 | NM_152811.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500023748.mm.1 | TC0500002690.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500023748.mm.1 | NM_152811.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500023748.mm.1 | NM_152811.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500023745.mm.1 | TC0500002690.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500023745.mm.1 | NM_152811.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500023745.mm.1 | NM_152811.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500023747.mm.1 | TC0500002690.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500023747.mm.1 | NM_152811.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500023747.mm.1 | NM_152811.1 | Refseq |
| Mouse Expression Set 430 | 1424934_PM_at | - | - |
| Mouse Expression Set 430 | 1424934_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424934_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424934_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424934_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424934_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 698898 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6650097 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6650097 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006650097 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1228666 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1228666 | - | - |
| mouse-6 v1.1 (Illumina) | scl26422.6_429-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6650097 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl26422.6_429-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6650097 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1228666 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1228666 | - | - |

Ugt2b1
Genomic Map