Assay Details
| Gene Symbol: Plcd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152813.3 | NP_690026.2 | 8 - 9 | 1526 | 102 | |
| GenBank mRNA | AK011892.1 | - | 4 - 5 | 457 | 102 | |
| AK046669.1 | - | 8 - 9 | 1492 | 102 | ||
| AK144950.1 | - | 2599 | 102 | |||
| BC031392.1 | - | 8 - 9 | 1470 | 4952407 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Plcd3
Genomic Map