Assay Details
| Gene Symbol: Entpd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026214.2 | NP_001021385.1 | 11 - 12 | 1054 | 61 | |
| NM_001286049.1 | NP_001272978.1 | 10 - 11 | 1029 | 61 | ||
| NM_001286058.1 | NP_001272987.1 | 9 - 10 | 956 | 61 | ||
| NM_007647.3 | NP_031673.2 | 10 - 11 | 981 | 61 | ||
| GenBank mRNA | AF006482.1 | - | 10 - 11 | 989 | 61 | |
| AF136571.1 | - | 10 - 11 | 1015 | 61 | ||
| AJ238636.1 | - | 10 - 11 | 1017 | 61 | ||
| AK002618.1 | - | 10 - 11 | 954 | 61 | ||
| AK031581.1 | - | 11 - 12 | 1034 | 61 | ||
| AK036641.1 | - | 11 - 12 | 1031 | 61 | ||
| AK045828.1 | - | 10 - 11 | 967 | 61 | ||
| AK079267.1 | - | 9 - 10 | 878 | 61 | ||
| AK080265.1 | - | 11 - 12 | 1041 | 61 | ||
| AK081435.1 | - | 11 - 12 | 1019 | 61 | ||
| AK088455.1 | - | 10 - 11 | 1010 | 61 | ||
| AK160950.1 | - | 8 - 9 | 875 | 61 | ||
| AK163688.1 | - | 10 - 11 | 955 | 61 | ||
| BC015247.1 | - | 9 - 10 | 939 | 4242560 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Entpd5
Genomic Map