Assay Details
| Gene Symbol: Ctsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007798.3 | NP_031824.1 | 1 - 2 | 107 | 100 | |
| GenBank mRNA | AK002456.1 | - | 1 - 2 | 56 | 99 | |
| AK083393.1 | - | 1 - 2 | 107 | 100 | ||
| AK159992.1 | - | 1 - 2 | 92 | 100 | ||
| AK167361.1 | - | 1 - 2 | 88 | 100 | ||
| AK170959.1 | - | 1 - 2 | 80 | 100 | ||
| BC006656.1 | - | 1 - 2 | 98 | 3500700 | 100 | |
| S69034.1 | - | 1 - 2 | 103 | 100 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK083393.1,AK159992.1,AK167361.1,AK170959.1,BC006656.1,NM_007798.3,S69034.1 transcripts.
Set Membership: |
|

Ctsb
Genomic Map