Assay Details
| Gene Symbol: Ctsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007798.3 | NP_031824.1 | 4 - 5 | 459 | 82 | |
| GenBank mRNA | AK002456.1 | - | 4 - 5 | 408 | 82 | |
| AK083393.1 | - | 4 - 5 | 459 | 82 | ||
| AK147192.1 | - | 4 - 5 | 406 | 82 | ||
| AK149884.1 | - | 4 - 5 | 408 | 82 | ||
| AK151790.1 | - | 4 - 5 | 406 | 82 | ||
| AK153047.1 | - | 4 - 5 | 405 | 82 | ||
| AK159992.1 | - | 4 - 5 | 444 | 82 | ||
| AK167361.1 | - | 4 - 5 | 440 | 82 | ||
| AK170959.1 | - | 4 - 5 | 432 | 82 | ||
| BC006656.1 | - | 4 - 5 | 450 | 3500700 | 82 | |
| M14222.1 | - | 3 - 4 | 364 | 82 | ||
| S69034.1 | - | 4 - 5 | 455 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ctsb
Genomic Map