| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900009054.mm.1 |
NM_007801.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900009054.mm.1 |
TC0900001155.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900009060.mm.1 |
NM_007801.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900009060.mm.1 |
TC0900001155.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900009059.mm.1 |
NM_007801.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900009059.mm.1 |
TC0900001155.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900009062.mm.1 |
NM_007801.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900009062.mm.1 |
TC0900001155.mm.1 |
MTA TC |