Assay Details
| Gene Symbol: Cttn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252572.1 | NP_001239501.1 | 2 - 3 | 333 | 80 | |
| NM_007803.5 | NP_031829.2 | 2 - 3 | 333 | 80 | ||
| GenBank mRNA | AK084249.1 | - | 2 - 3 | 143 | 80 | |
| AK146856.1 | - | 2 - 3 | 140 | 80 | ||
| AK148010.1 | - | 2 - 3 | 136 | 80 | ||
| AK165555.1 | - | 2 - 3 | 136 | 80 | ||
| AK168699.1 | - | 2 - 3 | 134 | 80 | ||
| AK168871.1 | - | 2 - 3 | 134 | 80 | ||
| AK168945.1 | - | 2 - 3 | 144 | 80 | ||
| AK196419.1 | - | 1 - 2 | 88 | 80 | ||
| AK196614.1 | - | 1 - 2 | 88 | 80 | ||
| AK202001.1 | - | 1 - 2 | 88 | 80 | ||
| AK203816.1 | - | 1 - 2 | 88 | 80 | ||
| AK218098.1 | - | 1 - 2 | 88 | 80 | ||
| BC011434.1 | - | 2 - 3 | 126 | 3981559 | 80 | |
| U03184.1 | - | 2 - 3 | 122 | 80 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK084249.1,AK146856.1,AK148010.1,AK165555.1,AK168699.1,AK168945.1,BC011434.1,NM_001252572.1,NM_007803.5,U03184.1 transcripts.
Set Membership: |
|

Cttn
Genomic Map